Learn More
Biomatik Corporation Human Peroxisome Proliferators Activator Receptors Alpha, Ppar-Α Elisa Kit
Supplier:  Biomatik Corporation EKC40684
Encompass_Preferred
Human Peroxisome Proliferators Activator Receptors Alpha, Ppar-Α Elisa Kit for detecting Peroxisome Proliferator-Activated Receptor Alpha in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q07869; PPARA. Target Synonyms: hPPAR; MGC2237; MGC2452; NR1C1; Nuclear receptor subfamily 1 group C member 1; OTTHUMP00000197740; OTTHUMP00000197741; Peroxisome proliferative activated receptor alpha; Peroxisome proliferator activated receptor alpha; Peroxisome proliferator-activated receptor alpha; PPAR; PPAR-alpha; ppara; PPARA_HUMAN; PPARalpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4779