Learn More
Biomatik Corporation Human Kallikrein 10,KLK 10 ELISA Kit
Supplier:  Biomatik Corporation EKC40744
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 1.675 ng/mL-40 ng/mL. Sensitivity: 0.7 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O43240. Target Synonyms: NES1, PRSSL1, breast normal epithelial cell associated serine protease|kallikrein 10|normal epithelial cell-specific 1|protease, serine-like, 1. Restriction: For Research Use Only.
Catalog No. 50-229-4791