Learn More
Biomatik Corporation Dog Interleukin 2, Il-2 Elisa Kit
Supplier:  Biomatik Corporation EKC40790
Encompass_Preferred
Dog Interleukin 2, Il-2 Elisa Kit for detecting Interleukin 2 in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q29416; IL2. Target Synonyms: IL2Interleukin-2; IL-2; T-cell growth factor; TCGF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4796