Learn More
Biomatik Corporation Human Regenerating Gene Iv, Reg-4 Elisa Kit
Supplier:  Biomatik Corporation EKC40824
Encompass_Preferred
Human Regenerating Gene Iv, Reg-4 Elisa Kit for detecting Regenerating Islet-Derived Family, Member 4 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9BYZ8; REG4. Target Synonyms: Gastrointestinal secretory protein; GISP; Reg IV; REG like protein; REG-4; REG-like protein; Reg4; REG4_HUMAN; Regenerating gene type IV; Regenerating islet derived family member 4; Regenerating islet derived protein 4 precursor; Regenerating islet-derived protein 4; Regenerating islet-derived protein IV; REGIV; RELP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4802