Learn More
Biomatik Corporation Rat Fatty Acid Binding Protein 4, Adipocyte (Fabp4) Elisa Kit
Supplier:  Biomatik Corporation EKC40919
Encompass_Preferred
Rat Fatty Acid Binding Protein 4, Adipocyte (Fabp4) Elisa Kit for detecting Fatty Acid Binding Protein 4, Adipocyte in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P70623; Fabp4. Target Synonyms: Fabp4Fatty acid-binding protein; adipocyte; Adipocyte lipid-binding protein; ALBP; Adipocyte-type fatty acid-binding protein; A-FABP; AFABP; Fatty acid-binding protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4809