Learn More
Biomatik Corporation Dog Osteocalcin/Bone Gla Protein, Ot/Bgp Elisa Kit
Supplier:  Biomatik Corporation EKC40986
Encompass_Preferred
Dog Osteocalcin/Bone Gla Protein, Ot/Bgp Elisa Kit for detecting Bone Gamma-Carboxyglutamate (Gla) Protein in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P81455; BGLAP. Target Synonyms: BGLAP; Osteocalcin; Bone Gla protein; BGP; Gamma-carboxyglutamic acid-containing protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4816