Learn More
Biomatik Corporation Rat Xanthine Oxidase(XO) ELISA Kit
Supplier:  Biomatik Corporation EKC41034
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.312 ng/mL-20 ng/mL. Sensitivity: 0.078 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P22985. Target Synonyms: XO, XOR,XDH, xanthene dehydrogenase|xanthine oxidase|xanthine oxidoreductase. Restriction: For Research Use Only.
Catalog No. 50-229-4825