Learn More
Biomatik Corporation Human Fibroblast Growth Factor-13, Fgf-13 Elisa Kit
Supplier:  Biomatik Corporation EKC41068
Encompass_Preferred
Human Fibroblast Growth Factor-13, Fgf-13 Elisa Kit for detecting Fibroblast Growth Factor 13 in serum, plasma, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 12.5-800pg/ml. Sensitivity: 3.12pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q92913; FGF13. Target Synonyms: FGF 13; FGF 2; FGF-13; FGF13; FGF13_HUMAN; FGF2; FHF 2; FHF-2; FHF2; Fibroblast growth factor 13; Fibroblast growth factor homologous factor 2; OTTHUMP00000024143; OTTHUMP00000024144. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4833