Learn More
Biomatik Corporation Human Tumor Necrosis Factor Receptor-Associated Factor 6, Traf6 Elisa Kit
Supplier:  Biomatik Corporation EKC41102
Encompass_Preferred
Human Tumor Necrosis Factor Receptor-Associated Factor 6, Traf6 Elisa Kit for detecting Tnf Receptor-Associated Factor 6 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 39-2500pg/ml. Sensitivity: 9.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9Y4K3; TRAF6. Target Synonyms: E3 ubiquitin-protein ligase TRAF6; Interleukin 1 signal transducer; Interleukin-1 signal transducer; MGC 3310; MGC:3310; MGC3310; OTTHUMP00000232772; OTTHUMP00000232773; RING finger protein 85; RNF 85; RNF85; TNF receptor associated factor 6; TNF receptor-associated factor 6; TNF receptor-associated factor 6; E3 ubiquitin protein ligase; TRAF 6; Traf6; TRAF6_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4838