Learn More
Biomatik Corporation Rabbit Matrix Metalloproteinase 13,MMP-13 ELISA Kit
Supplier:  Biomatik Corporation EKC41127
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 93.75 pg/mL-6000 pg/mL. Sensitivity: 23.44 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O62806. Target Synonyms: CLG3, MANDP1, collagenase 3|matrix metalloproteinase 13|matrix metalloproteinase 13 (collagenase 3). Restriction: For Research Use Only.
Catalog No. 50-229-4841