Learn More
Biomatik Corporation Sheep 5-hydroxytryptamine/serotonin(5HT/ST) ELISA kit
Supplier:  Biomatik Corporation EKC41155
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Ovis aries (Sheep). Detection Range: 0.625 ng/mL-10 ng/mL. Sensitivity: 0.312 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4845