Learn More
Biomatik Corporation Human Soluble Tumor Necrosis Factor-Like Weak Inducer Of Apoptosis (Stweak) Elisa Kit
Supplier:  Biomatik Corporation EKC41226
Encompass_Preferred
Human Soluble Tumor Necrosis Factor-Like Weak Inducer Of Apoptosis (Stweak) Elisa Kit for detecting Tumor Necrosis Factor (Ligand) Superfamily, Member 12 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O43508; TNFSF12. Target Synonyms: APO 3 ligand; APO 3L; APO3 ligand; APO3/DR3 ligand; APO3L; DR3LG; MGC129581; MGC20669; secreted form; TNF-related weak inducer of apoptosis; TNF12_HUMAN; TNFSF 12; Tnfsf12; TNFSF12 protein; Tumor necrosis factor (ligand) superfamily member 12; Tumor necrosis factor ligand superfamily member 12; Tumor necrosis factor superfamily member 12; TWEAK; UNQ181/PRO207. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4853