Learn More
Biomatik Corporation Human Arachidonate 12-Lipoxygenase, 12S-Type (Alox12/Log12) Elisa Kit
Supplier:  Biomatik Corporation EKC41250
Encompass_Preferred
Human Arachidonate 12-Lipoxygenase, 12S-Type (Alox12/Log12) Elisa Kit for detecting Arachidonate 12-Lipoxygenase in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P18054; ALOX12. Target Synonyms: 12 LOX; 12 (S) lipoxygenase; 12-lipoxygenase; 12LO; 12S LOX; 12S-lipoxygenase; 12S-LOX; 12S-type; Alox12; Arachidonate 12 lipoxygenase; Arachidonate 12 lipoxygenase; 12S type; Arachidonate 12-lipoxygenase; Arachidonate 12-oxidoreductase; Lipoxin synthase 12-LO; LOG12; LOX12_HUMAN; P-12LO; Platelet 12-LOX; Platelet type lipoxygenase 12; platelet-type 12-lipoxygenase; Platelet-type lipoxygenase 12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4856