Learn More
Biomatik Corporation Human Transgelin (Tagln) Elisa Kit
Supplier:  Biomatik Corporation EKC41318
Encompass_Preferred
Human Transgelin (Tagln) Elisa Kit for detecting Transgelin in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q01995; TAGLN. Target Synonyms: TAGLN; SM22; WS3-10; Transgelin; 22 kDa actin-binding protein; Protein WS3-10; Smooth muscle protein 22-alpha; SM22-alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4861