Learn More
Biomatik Corporation Human Bmp And Activin Membrane-Bound Inhibitor Homolog (Bambi) Elisa Kit
Supplier:  Biomatik Corporation EKC41376
Encompass_Preferred
Human Bmp And Activin Membrane-Bound Inhibitor Homolog (Bambi) Elisa Kit for detecting Bmp And Activin Membrane-Bound Inhibitor Homolog (Xenopus Laevis) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q13145; BAMBI. Target Synonyms: BAMBI; BAMBI_HUMAN; BMP and activin membrane bound inhibitor homolog; BMP and activin membrane-bound inhibitor homolog; NMA; Non metastatic gene A protein; Non-metastatic gene A protein; Putative transmembrane protein NMA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4867