Learn More
Biomatik Corporation Human Bone Morphogenetic Protein 3 (Bmp3) Elisa Kit
Supplier:  Biomatik Corporation EKC41386
Encompass_Preferred
Human Bone Morphogenetic Protein 3 (Bmp3) Elisa Kit for detecting Bone Morphogenetic Protein 3 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P12645; BMP3. Target Synonyms: BMP-3; BMP-3A; bmp3; BMP3_HUMAN; BMP3A; Bone morphogenetic protein 3 (osteogenic); Bone morphogenetic protein 3; Bone morphogenetic protein 3A; Osteogenin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4869