Learn More
Biomatik Corporation Human Chordin-Like Protein 1 (Chrdl1) Elisa Kit
Supplier:  Biomatik Corporation EKC41450
Encompass_Preferred
Human Chordin-Like Protein 1 (Chrdl1) Elisa Kit for detecting Chordin-Like 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9BU40; CHRDL1. Target Synonyms: CHL; chordin like 1; Chordin-like protein 1; CHRDL1; CRDL1_HUMAN; dA141H5.1; neuralin 1; Neuralin-1; Neurogenesin-1; NRLN1; Ventroptin; VOPT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4874