Learn More
Biomatik Corporation Mouse Cardiotrophin-Like Cytokine Factor 1 (Clcf1) Elisa Kit
Supplier:  Biomatik Corporation EKC41456
Encompass_Preferred
Mouse Cardiotrophin-Like Cytokine Factor 1 (Clcf1) Elisa Kit for detecting Cardiotrophin-Like Cytokine Factor 1 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9QZM3; Clcf1. Target Synonyms: Clcf1; Bsf3; Nnt1Cardiotrophin-like cytokine factor 1; B-cell-stimulating factor 3; BSF-3; Novel neurotrophin-1; NNT-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4876