Learn More
Biomatik Corporation Mouse C-X-C motif chemokine 9(CXCL9) ELISA kit
Supplier:  Biomatik Corporation EKC41506
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P18340. Target Synonyms: CMK, Humig, MIG, SCYB9, crg-10, monokine induced by gamma interferon|small inducible cytokine B9. Restriction: For Research Use Only.
Catalog No. 50-229-4880