Learn More
Biomatik Corporation Human Fibroblast Growth Factor 19 (Fgf19) Elisa Kit
Supplier:  Biomatik Corporation EKC41569
Encompass_Preferred
Human Fibroblast Growth Factor 19 (Fgf19) Elisa Kit for detecting Fibroblast Growth Factor 19 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O95750; FGF19. Target Synonyms: FGF 19; FGF-19; FGF15; FGF19; FGF19_HUMAN; Fibroblast growth factor 15; Fibroblast growth factor 19. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4891