Learn More
Biomatik Corporation Dog Gastrin (Gast) Elisa Kit
Supplier:  Biomatik Corporation EKC41590
Encompass_Preferred
Dog Gastrin (Gast) Elisa Kit for detecting Gastrin in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01353; GAST. Target Synonyms: GAST; GASGastrin [Cleaved into: Big gastrin; Gastrin-34; G34); Gastrin]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4896