Learn More
Biomatik Corporation Human Growth Hormone Variant (Gh2) Elisa Kit
Supplier:  Biomatik Corporation EKC41600
Encompass_Preferred
Human Growth Hormone Variant (Gh2) Elisa Kit for detecting Growth Hormone Variant(Gh2) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 133-4000pg/ml. Sensitivity: 33pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01242; GH2. Target Synonyms: GH2; Growth hormone variant; GH-V; Growth hormone 2; Placenta-specific growth hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4898