Learn More
Biomatik Corporation Human Interleukin-1 Family Member 10 (Il1F10) Elisa Kit
Supplier:  Biomatik Corporation EKC41681
Encompass_Preferred
Human Interleukin-1 Family Member 10 (Il1F10) Elisa Kit for detecting Interleukin 1 Family, Member 10 (Theta) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8WWZ1; IL1F10. Target Synonyms: family of interleukin 1-theta; FIL1 theta; FIL1T; FKSG75; IL 38; IL-1 theta; IL-1F10 (canonical form IL-1F10a); IL-1F10; IL-1HY2; IL1 theta; IL1F10; IL1FA_HUMAN; IL1HY2; Interleukin 1 family member 10 (theta); Interleukin 1 receptor antagonist FKSG75; Interleukin 1 receptor antagonist homolog 2; Interleukin 1 receptor antagonist like FIL1 theta; Interleukin-1 family member 10; Interleukin-1 HY2; Interleukin-1 theta; interleukin-38. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4919