Learn More
Biomatik Corporation Human Protein Jagged-1 (Jag1) Elisa Kit
Supplier:  Biomatik Corporation EKC41715
Encompass_Preferred
Human Protein Jagged-1 (Jag1) Elisa Kit for detecting Jagged 1 (Alagille Syndrome) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P78504; JAG1. Target Synonyms: AGS; AHD; AWS; CD 339; CD339; CD339 antigen; Headturner; hJ1; Htu; Jag 1; Jag1; JAG1_HUMAN; Jagged 1; Jagged1 (Alagille syndrome); Jagged1; JAGL1; MGC104644; OTTHUMP00000030278; Protein jagged-1; Ser 1; Ser1; Serrate 1; Slalom. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4925