Learn More
Biomatik Corporation Human Kallikrein-5 (Klk5) Elisa Kit
Supplier:  Biomatik Corporation EKC41723
Encompass_Preferred
Human Kallikrein-5 (Klk5) Elisa Kit for detecting Kallikrein-Related Peptidase 5 in serum, plasma, cell culture supernates, tissue homogenates, saliva. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9Y337; KLK5. Target Synonyms: HK5; Kallikrein 5; Kallikrein L2; Kallikrein like 2; Kallikrein like protein 2; Kallikrein related peptidase 5; Kallikrein-5; Kallikrein-like protein 2; Kallikrein5; KLK 5; KLK L2; KLK-L2; KLK5; KLK5_HUMAN; KLKL 2; KLKL2; SCTE; Stratum corneum tryptic enzyme; UNQ570/PRO1132. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4928