Learn More
Biomatik Corporation Mouse Lactadherin(MFGE8) ELISA kit
Supplier:  Biomatik Corporation EKC41776
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.8 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P21956. Target Synonyms: BA46, EDIL1, HsT19888, MFG-E8, OAcGD3S, SED1, SPAG10, hP47, O-acetyl disialoganglioside synthase|lactadherin|medin|sperm associated antigen 10|sperm surface protein hP47. Restriction: For Research Use Only.
Catalog No. 50-229-4937