Learn More
Biomatik Corporation Human Methionine Synthase (Mtr) Elisa Kit
Supplier:  Biomatik Corporation EKC41792
Encompass_Preferred
Human Methionine Synthase (Mtr) Elisa Kit for detecting 5-Methyltetrahydrofolate-Homocysteine Methyltransferase in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q99707; MTR. Target Synonyms: MTRMethionine synthase; EC 2.1.1.13; 5-methyltetrahydrofolate--homocysteine methyltransferase; Vitamin-B12 dependent methionine synthase; MS. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4939