Learn More
Biomatik Corporation Rat Netrin-1(NTN1) ELISA kit
Supplier:  Biomatik Corporation EKC41817
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 18.75 pg/mL-1200 pg/mL. Sensitivity: 4.7 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q924Z9. Target Synonyms: NTN1L, Netrin-1|netrin 1, mouse, homolog of. Restriction: For Research Use Only.
Catalog No. 50-229-4945