Learn More
Biomatik Corporation Human Peptidase Inhibitor 16 (Pi16) Elisa Kit
Supplier:  Biomatik Corporation EKC41832
Encompass_Preferred
Human Peptidase Inhibitor 16 (Pi16) Elisa Kit for detecting Peptidase Inhibitor 16 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-10ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q6UXB8; PI16. Target Synonyms: Cripi; CRISP 9; CRISP-9; CRISP9; Cysteine rich protease inhibitor; Cysteine rich secretory protein 9; Cysteine-rich secretory protein 9; Microseminoprotein beta binding protein; MSMBBP; Peptidase inhibitor 16; PI-16; PI16; PI16_HUMAN; Protease inhibitor 16; PSP94 binding protein; PSP94-binding protein; PSPBP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4949