Learn More
Biomatik Corporation Human Phospholipase A1 Member A (Pla1A) Elisa Kit
Supplier:  Biomatik Corporation EKC41836
Encompass_Preferred
Human Phospholipase A1 Member A (Pla1A) Elisa Kit for detecting Phospholipase A1 Member A in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q53H76; PLA1A. Target Synonyms: AA986889; NMD; OTTHUMP00000215248; OTTHUMP00000215249; OTTHUMP00000215250; Phosphatidylserine specific phospholipase A1; Phosphatidylserine specific phospholipase A1alpha; Phosphatidylserine-specific phospholipase A1; Phospholipase A1 member A; pla1a; PLA1A_HUMAN; PS PLA1; PS-PLA1; PSPLA 1; PSPLA1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4951