Learn More
Biomatik Corporation Human Roundabout Homolog 1 (Robo1) Elisa Kit
Supplier:  Biomatik Corporation EKC41884
Encompass_Preferred
Human Roundabout Homolog 1 (Robo1) Elisa Kit for detecting Roundabout, Axon Guidance Receptor, Homolog 1 (Drosophila) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9Y6N7; ROBO1. Target Synonyms: Deleted in U twenty twenty; DUTT 1; DUTT1; FLJ21882; H Robo 1; H-Robo-1; hRobo 1; Robo 1; Robo1; ROBO1_HUMAN; Roundabout 1; Roundabout axon guidance receptor homolog 1; Roundabout homolog 1; Roundabout homolog1 precurser; Roundabout1; SAX 3; SAX3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4959