Learn More
Biomatik Corporation Human Protein S100-A10 (S100A10) Elisa Kit
Supplier:  Biomatik Corporation EKC41889
Encompass_Preferred
Human Protein S100-A10 (S100A10) Elisa Kit for detecting S100 Calcium Binding Protein A10 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P60903; S100A10. Target Synonyms: 42C; AA409961; AL024248; Annexin II ligand; Annexin II ligand; calpactin I; light polypeptide; Annexin II tetramer (AIIt) p11 subunit; Annexin II; light chain; ANX2L; ANX2LG; Ca[1]; CAL12; CAL1L; Calpactin I light chain; Calpactin I; p11 subunit; Calpactin-1 light chain; Cellular ligand of annexin II; CLP11; GP11; MGC111133; Nerve growth factor-induced protein 42C; OTTHUMP00000015269; OTTHUMP00000015270; p10; p10 protein; p11; Protein S100 A10; Protein S100-A10; S100 calcium binding protein A10 (annexin II ligand; calpactin I; light polypeptide (p11)). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4962