Learn More
Biomatik Corporation Human Transgelin-2 (Tagln2) Elisa Kit
Supplier:  Biomatik Corporation EKC41955
Encompass_Preferred
Human Transgelin-2 (Tagln2) Elisa Kit for detecting Transgelin 2 in serum, plasma, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P37802; TAGLN2. Target Synonyms: CDABP0035; epididymis tissue protein Li 7e; HA1756; KIAA0120; SM22-alpha homolog; TAGL2_HUMAN; Tagln2; Transgelin-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4972