Learn More
Biomatik Corporation Human Tumor Necrosis Factor Ligand Superfamily Member 9 (Tnfsf9) Elisa Kit
Supplier:  Biomatik Corporation EKC41991
Encompass_Preferred
Human Tumor Necrosis Factor Ligand Superfamily Member 9 (Tnfsf9) Elisa Kit for detecting Tumor Necrosis Factor (Ligand) Superfamily, Member 9 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 18.75-1200pg/ml. Sensitivity: 4.7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P41273; TNFSF9. Target Synonyms: 4 1BB L; 4 1BB ligand; 4 1BBL; 4-1BB ligand; 4-1BBL; Cd137l; Cd157l; Homolog of mouse 4 1BB L; Homolog of mouse 4 1BBL; ILA ligand (TNF related); Ly63l; Receptor 4 1BB ligand; TNF superfamily member 9; TNFL9_HUMAN; Tnfsf9; TNLG5A; Tumor necrosis factor (ligand) superfamily member 9; Tumor necrosis factor ligand 5A; Tumor necrosis factor ligand superfamily member 9; Tumor necrosis factor superfamily member 9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4977