Learn More
Biomatik Corporation Human Pantetheinase (Vnn1) Elisa Kit
Supplier:  Biomatik Corporation EKC42011
Encompass_Preferred
Human Pantetheinase (Vnn1) Elisa Kit for detecting Vanin 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O95497; VNN1. Target Synonyms: HDLCQ8; High density lipoprotein cholesterol level quantitative trait locus 8; included; Pantetheinase; Pantetheine hydrolase; Tiff66; V-1; Vanin 1; Vanin-1; Vannin 1; Vascular non inflammatory molecule 1; Vascular non-inflammatory molecule 1; VNN 1; VNN1; VNN1_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4980