Learn More
Biomatik Corporation Chicken Rudimental Bovine Serum Albumin (Bsa) Check-Up Elisa Kit
Supplier:  Biomatik Corporation EKC42048
Encompass_Preferred
Chicken Rudimental Bovine Serum Albumin (Bsa) Check-Up Elisa Kit for detecting Rudimental Bovine Serum Albumin(Bsa) Check-Up in serum, plasma, tissue homogenates. Species Reactivity: Chicken (Gallus). Detection Range: 0.312-80ug/ml. Sensitivity: 0.312ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4988