Learn More
Biomatik Corporation Guinea pig tri-iodothyronine (T3) ELISA kit
Supplier:  Biomatik Corporation EKC42069
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Cavia porcellus (Guinea pig). Detection Range: 0.5 ng/mL-8 ng/mL. Sensitivity: 0.5 ng/mL. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4990