Learn More
Biomatik Corporation Pig Free Tri-Iodothyronine (Ft3) Elisa Kit
Supplier:  Biomatik Corporation EKC42190
Encompass_Preferred
Pig Free Tri-Iodothyronine (Ft3) Elisa Kit for detecting Free Tri-Iodothyronine Indes(Ft3) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 2-32pmol/l. Sensitivity: 0.38pmol/l. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5000