Learn More
Biomatik Corporation Human beta cellulin,BTC ELISA Kit
Supplier:  Biomatik Corporation EKC42207
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P35070. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-5002