Learn More
Biomatik Corporation Human Fms-Like Tyrosine Kinase 3 Ligand, Flt-3L Elisa Kit
Supplier:  Biomatik Corporation EKC42210
Encompass_Preferred
Human Fms-Like Tyrosine Kinase 3 Ligand, Flt-3L Elisa Kit for detecting Fms-Related Tyrosine Kinase 3 Ligand in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P49771; FLT3LG. Target Synonyms: FL; Flt 3 ligand; Flt 3L; Flt3 L; FLT3 LG; Flt3 ligand; Flt3L; FLT3L_HUMAN; Flt3lg; Fms related tyrosine kinase 3 ligand; Fms-related tyrosine kinase 3 ligand; SL cytokine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5005