Learn More
Biomatik Corporation Mouse Interleukin 3,IL-3 ELISA KIT
Supplier:  Biomatik Corporation EKC42213
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 7.8 pg/mL-500 pg/mL. Sensitivity: 1.95 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P01586. Target Synonyms: IL-3, MCGF, MGC79398, MGC79399, MULTI-CSF, P-cell stimulating factor|hematopoietic growth factor|interleukin 3|mast-cell growth factor|multilineage-colony-stimulating factor. Restriction: For Research Use Only.
Catalog No. 50-229-5008