Learn More
Biomatik Corporation Human Squamous Cell Carcinoma Antigen 2,SCCAg 2 ELISA Kit
Supplier:  Biomatik Corporation EKC42226
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312 ng/ml-20 ng/ml. Sensitivity: 0.078 ng/ml. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P48594. Target Synonyms: LEUPIN, PI11, SCCA-2, SCCA1, SCCA2, SCCA2/SCCA1 fusion protein|protease inhibitor (leucine-serpin)|serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 4|squamous cell carcinoma an. Restriction: For Research Use Only.
Catalog No. 50-229-5016