Learn More
Biomatik Corporation Guinea Pig Matrix Metalloproteinase 9, Mmp-9 Elisa Kit
Supplier:  Biomatik Corporation EKC42228
Encompass_Preferred
Guinea Pig Matrix Metalloproteinase 9, Mmp-9 Elisa Kit for detecting Pig Matrix Metalloproteinase 9, Mmp-9 in serum, plasma, tissue homogenates. Species Reactivity: Guinea pig (Cavia porcellus). Detection Range: 3.12-200 ng/ ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: H0VAD9; MMP9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5018