Learn More
Biomatik Corporation Human Myeloid Progenitor Inhibitory Factor 1, Mpif-1 Elisa Kit
Supplier:  Biomatik Corporation EKC42233
Encompass_Preferred
Human Myeloid Progenitor Inhibitory Factor 1, Mpif-1 Elisa Kit for detecting Chemokine (C-C Motif) Ligand 23 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 39-2500pg/ml. Sensitivity: 9.7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P55773; CCL23. Target Synonyms: C C motif chemokine ligand 23; C-C motif chemokine 23; C6 beta chemokine; CCL 23; CCL23; CCL23 (30-99); CCL23_HUMAN; Chemokine (C C motif) ligand 23; Chemokine CC motif ligand 23; ck beta 8; CK-beta-8; CKB 8; CKB-8; Ckb-8-1; CKB8; Hmrp-2a; Macrophage inflammatory protein 3; MIP 3; MIP-3; MIP3; MPIF 1; MPIF-1; MPIF1; Myeloid progenitor inhibitory factor 1; OTTHUMP00000163948; SCYA 23; SCYA23; Small inducible cytokine A 23; Small inducible cytokine subfamily A (Cys-Cys); member 23; Small inducible cytokine subfamily A; member 23; Small-inducible cytokine A23. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5022