Learn More
Biomatik Corporation Rat Insulin-like growth factor binding protein 2,IGFBP-2 ELISA kit
Supplier:  Biomatik Corporation EKC42240
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.625 ng/mL-40 ng/mL. Sensitivity: 0.156 ng/mL. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P12843. Target Synonyms: IBP2, IGF-BP53. Restriction: For Research Use Only.
Catalog No. 50-229-5024