Learn More
Biomatik Corporation Human Heme Oxygenase 2, Ho-2 Elisa Kit
Supplier:  Biomatik Corporation EKC42261
Encompass_Preferred
Human Heme Oxygenase 2, Ho-2 Elisa Kit for detecting Heme Oxygenase (Decycling) 2 in serum, plasma, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P30519; HMOX2. Target Synonyms: Heme oxygenase (decycling) 2; Heme oxygenase (decyclizing) 2; Heme oxygenase 2; HMOX 2; Hmox2; HMOX2 protein; HMOX2_HUMAN; HO 2; HO-2; HO2; OTTHUMP00000159847. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5028