Learn More
Biomatik Corporation Human Cyclin-dependent kinase 4,CDK-4 ELISA Kit
Supplier:  Biomatik Corporation EKC42276
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P11802. Target Synonyms: CMM3, MGC14458, PSK-J3, cell division kinase 4|melanoma cutaneous malignant, 3. Restriction: For Research Use Only.
Catalog No. 50-229-5035