Learn More
Biomatik Corporation Mouse Soluble Endoglin,ENG/sCD105 ELISA Kit
Supplier:  Biomatik Corporation EKC42289
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 6.25 ng/mL-400 ng/mL. Sensitivity: 1.56 ng/mL. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q63961. Target Synonyms: RP11-228B15.2, CD105, END, FLJ41744, HHT1, ORW, ORW1, CD105 antigen. Restriction: For Research Use Only.
Catalog No. 50-229-5039