Learn More
Biomatik Corporation Human Granulocyte Chemotactic Protein-2, Gcp-2 Elisa Kit
Supplier:  Biomatik Corporation EKC42292
Encompass_Preferred
Human Granulocyte Chemotactic Protein-2, Gcp-2 Elisa Kit for detecting Chemokine (C-X-C Motif) Ligand 6 (Granulocyte Chemotactic Protein 2) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P80162; CXCL6. Target Synonyms: C X C motif chemokine 6; Chemokine (C X C motif) ligand 6; Chemokine alpha 3; CKA 3; CKA-3; CKA3; CXCL 6; CXCL6; CXCL6_HUMAN; GCP 2; GCP-2; Granulocyte chemotactic protein 2; N-processed variant 3; SCYB6; Small inducible cytokine B6; Small inducible cytokine subfamily B (Cys X Cys) member 6; Small inducible cytokine subfamily B (Cys X Cys) member b; Small-inducible cytokine B6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5041