Learn More
Biomatik Corporation Horse Interleukin 8, Il-8 Elisa Kit
Supplier:  Biomatik Corporation EKC42323
Encompass_Preferred
Horse Interleukin 8, Il-8 Elisa Kit for detecting Interleukin 8 in serum, plasma, tissue homogenates. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O62812; CXCL8. Target Synonyms: CXCL8; IL8; Interleukin-8; IL-8; C-X-C motif chemokine 8; Chemokine; C-X-C motif) ligand 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5051